Learn More
Sharpsguard™ orange com-plus Sharps Container
Designed to fit safely and securely into the Sharpsguard™ tray range and other carrying trays. Sharpsguard orange com-plus Sharps Container is for disposal of sharps that have not been contaminated with medicinal products.
Brand: Daniels Healthcare DD479OL
Description
- Rectangular for easy stowage in medical bags
- Sliding door with an aperture which will take a 20mL syringe
- Supplied with lid assembled
- Rigid polypropylene construction and wall thickness
- Offers outstanding protection against penetrating sharps and leaking fluids
- When container is full to the fill line it must be locked and disposed of safely
- All have clear British Standard markings, user instructions and a visible fill line
Specifications
0.78 L | |
Orange | |
205 mm | |
Autoclavable |
Lid | |
120 mm | |
54 x 36 mm (L x W) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.