missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Human IgA Secondary Antibody, Biotin, Invitrogen™
Sheep Polyclonal Secondary Antibody
Brand: Invitrogen PA516923
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PA5-16923 cross-reacts with bovine, mouse, and rat serum. The PA5-16923 immunogen is igA isolated from human serum.
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Human IgA | |
Polyclonal | |
TBS with 1% carrier protein and 0.1% sodium azide; pH 7.4 | |
IgA isolated from human serum | |
2 mL | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG |
Immunofluorescence | |
Biotin | |
Sheep | |
Purified | |
RUO | |
Human | |
Liquid |