Learn More
Sheep anti-Mouse Kappa Light Chain Secondary Antibody, FITC, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69223
Product Code. 18628878
  • 837.00 € / 1mL
Explore more special offers
Description
Description
Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
Specifications
Kappa Light Chain
Polyclonal
ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
A pool of purified Bence Jones kappa proteins isolated from mouse urine. Freund's complete adjuvant is used in the first step of the immunization procedure.
1 mL
Adaptive Immunity, Immunology
3514
Mouse
Lyophilized
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
FITC
Lyophilized from PBS (pH 7.2)
Sheep
Ion exchange chromatography
RUO
Secondary
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Store at 4°C in the dark.