Learn More
Sheep anti-Mouse Kappa Light Chain Secondary Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69222
Product Code. 18666697
  • 810.00 € / 10mg
Explore more special offers
Description
Description
Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoblot, Dot Blot
Specifications
Specifications
Kappa Light Chain
Polyclonal
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
A pool of purified Bence Jones kappa proteins isolated from mouse urine. Freund's complete adjuvant is used in the first step of the immunization procedure.
10 mg
Adaptive Immunity, Immunology
3514
Mouse
Lyophilized
Western Blot, ELISA, Immunofluorescence, Immunoblot, Dot Blot
Unconjugated
Lyophilized from PBS (pH 7.2)
Sheep
Ion exchange chromatography
RUO
Secondary
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.