Learn More
Sheet Steel Oxygen Cylinder Trolleys

POA
Specifications
Type | Oxygen Cylinder Trolley |
---|---|
Material | Sheet Steel |
For Use With (Application) | Ideal for use in hospitals |
For Use With (Equipment) | Carrying oxygen cylinders |
Height (Metric) | 1100 mm |
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
12945961
![]() |
N/A
TS180L |
470 x 480 x 1100 mm |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12935961
![]() |
N/A
TS179L |
460 x 410 x 1100 mm |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The tubular frame has a sheet steel base and retaining straps to hold the cylinder in position. Each trolley is fitted with anti-static metal centred wheels and front 100mm non-marking corner buffers.
Specifications
Oxygen Cylinder Trolley | |
Ideal for use in hospitals | |
1100 mm |
Sheet Steel | |
Carrying oxygen cylinders |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.