Learn More
Thermo Scientific™ Shelf Kits for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with shelf kits designed for Thermo Scientific™ Ultra-Low Temperature Freezers.
Brand:  Thermo Scientific™ SK500
 View more versions of this product
Product Code. 11366401
  • 321.60 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize storage space with Thermo Scientific™ Shelf Kits for Thermo Scientific™ Ultra-Low Temperature Freezers; designed for use with with Thermo Scientific™ Forma™ 8800, HERAfreeze™ HFU-T, Revco™ UxF, and TSU Series freezers.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Includes one shelf and clips
Shelf Kit
1
Forma 88500; TSU500; HFU500T; Revco UxF500
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products