Learn More
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
Brand:  Thermo Scientific™ RSK13SD5
 View more versions of this product
Product Code. 11329351
  • 735.66 € / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
Features
  • Stainless steel racks
  • Boxes
  • Cell dividers
Warranty varies by country. Please contact us for details.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100
Shelf Kit
Boxes (48/4 Racks)
HERAfreeze HFU B Series, Revco ExF and DxF Series (13 cu. ft. models)
4 sliding drawer racks and (48) 2 in. boxes with 100 cell dividers
Shelf Kit
Stainless Steel
4