Learn More
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
Brand: Thermo Scientific™ RSK17SD5
This item is not returnable.
View return policy
Description
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
Features- Stainless steel racks
- Boxes
- Cell dividers
Warranty varies by country. Please contact us for details.
Specifications
100 | |
Shelf Kit | |
Boxes (80/4 Racks) | |
5 | |
4 |
4 sliding drawer racks and (80) 2 in. boxes with 100 cell dividers | |
Shelf Kit | |
Stainless Steel | |
HERAfreeze HFU B Series, Revco ExF and DxF Series (17 cu. ft. models) |