missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Shelf for RI150-RI250

Product Code. 15678175 Shop All Thermo Scientific Products
Click to view available options
Shelf Area (English):
21.3 x 17.3 in.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15678175

Brand: Thermo Scientific™ RISHELF

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Description Shelf for RI150-RI250
Type Shelf
Shelf Area (English) 21.3 x 17.3 in.
Shelf Area (Metric) 540 x 440 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.