missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Shell Vial Kit, Amber Glass, PE plug

Product Code. p-7113970 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
2.3 mL
4 mL
Unit Size:
Pack of 100

Product Code. 11565914

Only null left
Add to basket
Edge
Add to basket

Recommended for HPLC usage.

  • Shell vials and the appropriate plugs can also be supplied as a 2in1 KIT
  • A handy and inexpensive vial/ losure combination for uncritical analyses
TRUSTED_SUSTAINABILITY

Specifications

Description Shell Vial
Color Amber
Diameter (Metric) Outer 11.6 mm
Material Glass
Volume (Metric) 2.3 mL
Height (Metric) 31.5 mm
Closure Size 11
Marking Spot No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.