missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Shelves for Heratherm™ Ovens and Incubators
Perforated and wire mesh shelves
Brand: Thermo Scientific™ 50127770
This item is not returnable.
View return policy
Description
Perforated and wire mesh shelves
Specifications
Heratherm General Protocol Microbiological Incubator 60l | |
(2) Shelf Supports | |
1 | |
14.8 in. | |
14.8 in. |
Shelf | |
Stainless Steel | |
12.9 in. | |
37.5 cm | |
37.5 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction