Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
For gas detection
Brand:  Dräger™ CH28201
 View more versions of this product
Product Code. 10388421
  • 129.00 € / Each
Explore more special offers
Description
Description
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Specifications
Hydrogen Sulfide, Sulfur Dioxide 0.2%/A
0.2 to 7 vol.%
10
32°F to 104°F
40mg/L
1+2
2 min.
0°C to 40°C