Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
For gas detection
Brand:  Dräger™ 8101211
 View more versions of this product
Product Code. 12951677
  • 118.00 € / Each
Explore more special offers
Description
Description
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Specifications
Hydrogen Sulfide 2%/A
2 to 40 vol.%
10
32°F to 104°F
3 to 20mg/L
1
1 min.
0°C to 40°C