missing translation for 'onlineSavingsMsg'
Learn More
SHOWA™ 460 Series PVC-Coated Cold Resistant Gloves Product Code.: 17891171
Add to basket

SHOWA™ 460 Series PVC-Coated Cold Resistant Gloves

Product Code. 17891171
X-Large
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy

Product Code. 17891171

Brand: SHOWA™ 46010XL

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Insulated gloves protect hands, and remain flexible in low temperatures. Rough-textured PVC coating protects against oils and chemicals, while also enhancing grip capabilities and tactile precision.

  • Gloves remain flexible in temperatures as low as -4°F (-20°C)
  • PVC coating protects against oils and chemicals
  • Rough grip enhances tactile precision when handling small, greasy components
  • Extended wrist protection from harmful substances
  • Reduced potential exposure to bacteria, viruses, and fungi

Specifications

Coated Area Full
Coating Material PVC
Color Orange
Exterior Finish Fully Textured
Hand Specific
Size X-Large
For Use With (Application) Chemical Spray/Treatment, Coating Preparation, and Painting
Glove Cuff Style Gauntlet
Liner Material Acrylic
Series 460
Length (English) 11.8 in.
Length (Metric) 30 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SHOWA™ 460 Series PVC-Coated Cold Resistant Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.