missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ SICCO™ Desiccator Wall

Desiccator Wall made of acrylic glass and aluminum with six separate compartments.

Brand:  Bohlender™ V1994-08

Product Code. 17177744

  • 18315.00 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Aluminum frame with panels made of acrylic glass
  • Aix compartments and six doors
  • Includes six shelves made of stainless steel, six trays and six hygrometers, and four casters
  • Usable with up to 8 shelves per compartment
  • Reinforced aluminum frame with acrylic panels
  • All doors with magnetic catch and circular rubber seal
  • Four casters (two of the casters with brakes)
  • Ground clearance for comfortable handling
  • Easy to read electronic hygrometer
  • Variable height shelves made of stainless steel
  • Telescopic shelves
  • Ground clearance of lowest chamber: 240 mm
Specifications

Specifications

Acrylic
156 L
1080 mm
1870 mm
580 mm
100 kg
41.2 gal.
43 in.
74 in.
23 in.
220 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers