Learn More
Bohlender™ SICCO™ Extractor Hood
Extractor Hood Panels made of polycarbonate with a top and integrated connector DN 160 made of polypropylene.
Brand:  Bohlender™ V1960-04
 View more versions of this product
Product Code. 17117744
  • 4015.00 € / Each
Explore more special offers
Description
Description
  • Aluminum frame with panels made of polycarbonate
  • Top panel with integrated connector for exhaust system made of polypropylene
  • Robust and light-weight aluminum construction
  • Transparent and shatter-proof panels made of 5 mm thick polycarbonate
  • Standard connection DN 160 for aspirator tube
  • Superb all-round visibility
  • Large front-opening for comfortable working
  • Top side made of polypropylene for a good chemical resistance
Specifications
Specifications
Aluminum, Polycarbonate, Polypropylene
22 in.
560 mm
35 in.
900 mm
51 lb.
47 in.
1200 mm
24 in.
600 mm
DN 160 Threaded Necks
34.6 in.
880 mm
23 kg
46 in.
1160 mm