missing translation for 'onlineSavingsMsg'
Learn More

SID 7969543, Tocris Bioscience™

Product Code. 18074932
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective steroidogenic factor-1 (SF-1, NR5A1) inhibitor

Selective steroidogenic factor-1 (SF-1, NR5A1) inhibitor (IC50 values are 0.76, >33 and >33 μM at SF-1, RORα and VP16 respectively). Inhibits SF-1-dependent luciferase expression in HEK 293T cells in vitro (IC50 = 30 nM).

Chemical Identifiers

CAS 868224-64-0
Molecular Formula C24H24N2O7
Molecular Weight (g/mol) 452.463
InChI Key KWMBIIQCLUIHDI-UHFFFAOYSA-N
Synonym ethyl 2-2-2-2,3-dihydrobenzo b 1,4 dioxin-6-yl amino-2-oxoethyl-1-oxo-1,2-dihydroisoquinolin-5-yl oxy propanoate, ethyl 2-2-2-2,3-dihydro-1,4-benzodioxin-6-yl amino-2-oxoethyl-1,2-dihydro-1-oxo-5-isoquinolinyl oxy propanoate, isoquinolinone scaffold, 2, d0aj5g, 2-2-2,3-dihydrobenzo 1,4 dioxin-6-ylcarbamoyl methyl-1-oxo-1,2-dihydroisoquinolin-5-yloxy propionic acid ethyl ester, sid hplc , solid, ethyl 2-2-2,3-dihydro-1,4-benzodioxin-6-ylcarbamoyl methyl-1-oxo-1,2-dihydroisoquinolin-5-yl oxy propanoate, ethyl 2-2-2,3-dihydro-1,4-benzodioxin-6-yl carbamoyl methyl-1-oxo-1,2-dihydroisoquinolin-5-yl oxy propanoate, ethyl 2-2-2,3-dihydro-1,4-benzodioxin-6-yl carbamoyl methyl-1-oxoisoquinolin-5-yl oxy propanoate, ethyl 2-2-2-2,3-dihydro-benzo b 1,4 dioxine-6-yl amino-2-oxoethyl-1,2-dihydro-1-oxo-5-isoquinolinyl oxy propanoate
PubChem CID 4076092
IUPAC Name ethyl 2-[2-[2-(2,3-dihydro-1,4-benzodioxin-6-ylamino)-2-oxoethyl]-1-oxoisoquinolin-5-yl]oxypropanoate
SMILES CCOC(=O)C(C)OC1=CC=CC2=C1C=CN(C2=O)CC(=O)NC3=CC4=C(C=C3)OCCO4

Specifications

Quantity 10 mg
Formula Weight 452.46
Percent Purity >98%
Chemical Name or Material SID 7969543
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'