Learn More
Wesbart™ Side Access Freezer Rack
Wesbart side loading racks to store cryoboxes.
Brand: Wesbart™ SOR196
This item is not returnable.
View return policy
Description
ULT freezer racks for upright freezers to store boxesSpecifications
Freezer Rack | |
Autoclavable | |
Boxes (16) | |
Yes | |
560 x 140 x 235 mm |
Aluminum Side Access Rack | |
Silver | |
Rectangular | |
Non-sterile | |
235 mm |