Learn More
Wesbart™ Side Access Freezer Rack
Wesbart side loading racks to store cryoboxes.
Brand:  Wesbart™ SOR033
 View more versions of this product
Product Code. 17226914
  • 172.00 € / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
ULT freezer racks for upright freezers to store boxes
Specifications
Specifications
Freezer Rack
Autoclavable
Rectangular
Non-sterile
339 mm
Aluminum Side Access Rack
Boxes (24)
Yes
560 x 140 x 339 mm