Learn More
Scotch-Brite™ Sienna Diamond Floor Pads
For cleaning and conditioning worn or dull marble, terrazzo, slate, and polished concrete flooring. When floor appearance reaches a medium shine, the Scotch Brite™ Purple Diamond Floor Pad Plus can be used to further enhance the floor's appearance.
Brand:  Scotch-Brite™ 7100200175
 View more versions of this product
Product Code. 17128860
  • 214.00 € / Pack of 5
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Simultaneously cleans and polishes flooring to a brilliant gloss finish without the use of chemicals
  • Ideal for worn or dull marble, terrazzo, vinyl, slate, and polished concrete floors
  • Easy to use with no expensive or harmful chemicals needed, only water
  • Recommended for use with low and standard speed machines
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Floor Pad
406 mm
Low and Standard Speed Machines
Brown
Burnishing
Round