missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sieve Brush

A double-ended brush for use with Fisherbrand™ Stainless Steel Test Sieves.

Brand:  Fisherbrand™ FB70065-SV039

 View more versions of this product

Product Code. 11502763

  • 17.35 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Provides gentle brushing
  • Manufactured from brass with nylon bristles
  • Two different sizes: 30 x 135mm and 20 x 230mm
Specifications

Specifications

Term Brush
Brass
20 x 230 mm
Nylon
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.