missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Signal Waist Coat 8960-Yellow 100%PES
Shop All Uvex Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
missing translation for 'unitSize'
Each
Beschreibung
- Material: 100% polyester / mesh Safety vest made of polyester / mesh with space for logo attachment.
- Velcro fastener, 2 circumferential reflective stripes in the torso area, reflective strips over the shoulders for better visibility in stooped position, edging edges, logo application possible (position: back)
- Certified according to EN 20471 X2
- Size: M (50/52) - XXXL (66/68)
- Care instructions: 40° C washable, gentle cycle (max. 25 washes), chlorinate impossible, ironing impossible, dry cleaning: P (max. 20 cleaning cycles), tumble drying impossible
Spezifikation
Spezifikation
| Closure Type | Hook and Loop |
| Product Type | High-visibility Vest |
| Color | Yellow |
| Size | Medium |
| Material | Polyester |
| Thickness (Metric) | 140 g/m² |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur