Learn More
Epredia™ Signature Series™ Clarifier™ 1, 2
Enhance the nuclear definition and clarity of Signature Series hematoxylins with Epredia™ Signature Series Clarifier™ 1, 2.
18.45 € - 148.00 €
Product Code | Brand | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Type | Price | Quantity & Availability | ||||
10096348
|
Epredia™
7442 |
4 x 1 gal. | Clarifier 2 |
148.00 €
Pack of 4 |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||
10693629
|
Epredia™
7401 |
1 gal. | Clarifier 1 |
34.85 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10603639
|
Epredia™
7402 |
1 gal. | Clarifier 2 |
18.45 €
Each |
In Stock
Log in to see stock. |
||||
Description
- Improves contrast between Nuclei and Cytoplasm
- Increases cell transparency
- Removes background staining caused by excessive tissue adhesive
- Removes unwanted hematoxylin staining from adhesive without affecting nuclear staining
- Clarifier 1 is recommended with Hematoxylin 7211 and Hematoxylin 1
- Clarifier 2 is recommended with Hematoxylin 7212 and Hematoxylin 2
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362-Take off contaminated clothing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : acetic acid