missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Signature Series™ Hematoxylin 2

Yield visually similar results to Gills 2 and 3 with this uniquely formulated product.

Brand:  Epredia™ 7231

 View more versions of this product

Product Code. 10066478

  • 51.00 € / Pack of 4
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • No filtration required
  • Stains up to 2500 slides per pint
  • Part of the Signature Series Staining System
  • Crisp, well-defined nuclear detail chromatin delineation

Available in 5L packaging

  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging

Specifications

Specifications

Hematoxylin 2
1 pt.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers