missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Signature Series™ Hematoxylin 7211
Yield results that exceed those of Gill or Harris Hematoxylins with this uniquely formulated product.
Brand: Epredia™ 7211
40.90 EUR valid until 2025-12-31
Use promo code "24374" to get your promotional price.
This item is not returnable.
View return policy
Description
- No filtration required
- Stains up to 2500 slides per pint
- Crisp, well-defined nuclear detail
Available in 5 liter packaging
- Reduces plastic by 90%
- Increases stain stability
- Reduces messy stain spills
- Saves 35% storage space versus traditional packaging
Specifications
Hematoxylin | |
1 pt. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hematoxylin 7211
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction