missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ SILAC Advanced DMEM/F-12 Flex Media, no glucose, no phenol red
GREENER_CHOICE

SILAC Advanced DMEM/F-12 Flex Media, no glucose, no phenol red

Brand:  Gibco™ A2494301

71.25 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 15396278

  • 78.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

SILAC Advanced DMEM/F-12 Flex is Advanced DMEM/F-12 without L-arginine, L-glutamine, and L-lysine for use during SILAC protein labeling with stable isotopic labeled lysine and/or arginine. Advanced DMEM/F-12 is a widely used basal medium for supporting the growth of many different mammalian cells, which reduces the FBS requirements by 50-90% without any changes in growth rate or morphology. When supplemented with 1-2 % FBS, Advanced DMEM/F-12 Flex is capable of supporting cellular proliferation and maximum cell densities comparable to the conventional basal formulation supplemented with 5-10% FBS. Cells successfully cultured in Advanced DMEM/F-12 with no adaptation include Jurkat, WI-38, SP2, Vero, and MRC-5. We offer a variety of DMEM/F-12 modifications for a range of applications. Find the right formulation using the media selector tool. Advanced DMEM/F-12 is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride. Advanced DMEM/F-12 typically requires supplementation with 1-5% Fetal Bovine Serum and 4 mM L-glutamine or GlutaMAX™ supplement (optional). SILAC protein labeling experiments should be conducted using Gibco™ Dialyzed FBS. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced DMEM/F-12 uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5-10% CO2 environment to maintain physiological pH.

cGMP Manufacturing and Quality System
SILAC Advanced DMEM/F-12 Flex is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C) and protect from light.
Shelf life: 12 months from date of manufacture.
Mammalian Cell Culture
Dulbecco's Modified Eagle Medium
Sodium Pyruvate, Non Essential Amino Acids (NEAA)
Gibco™
Mammalian Cells
Liquid
Reduced Serum
No Glucose, No Glutamine, No HEPES, No Phenol Red
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.