missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ SILBERBRAND Automatic Burette with Amber Stopcock

SILBERBRAND Automatic Burette equipped with an amber lateral stopcock and PTFE key.

Brand:  BRAND™ 22316

229.09 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.


 View more versions of this product

Product Code. 10132111

  • 292.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Error limits according to Class B, DIN EN ISO 385
  • Interchangeable keys
  • Calibrated to deliver (TD, Ex)
  • With glass key in the titration stopcock
TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 mL
Without bottle
12 months
0.1 mL
Amber lateral stopcock with PTFE key
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ SILBERBRAND Automatic Burette with Amber Stopcock > Capacity: 25 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.