missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ SILBERBRAND Compact Automatic Burette
Compact Automatic Burette designed to be fast to dismantle and easy to clean.
Brand: BRAND™ 23830
153.05 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Easy to repair
- All individual components are replaceable
- Detachable PTFE stopcock
- Calibrated to deliver (TD, Ex)
- Error limit according to class B, DIN EN ISO 385
Specifications
±0.1 mL | |
50 mL | |
0.1 mL | |
12 months | |
Class B |
Amber | |
Borosilicate Glass (5.4) | |
1000 mL LDPE Bottle | |
Automatic Buret |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction