Learn More
BRAND™ SILBERBRAND Compact Burette with Schellbach Stripe
Compact Burette designed to be fast to dismantle and easy to clean.
Brand:  BRAND™ 13906
 View more versions of this product
Product Code. 10552063
  • 74.25 € / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With removable PTFE stopcock
  • All individual components are replaceable
  • Error limits according to class B, DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mL
Borosilicate Glass (3.3)
4 cm
PTFE
46.5 cm
0.1 mL
Black Schellbach Stripe, Stopcock
Buret