Learn More
Ambion™ Silencer™ Select Pre-Designed siRNA
Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.
149.00 € - 15330.00 €
Specifications
Description | Silencer™ Select Pre-Designed siRNA |
---|---|
RNAi Type | siRNA |
Product Line | Silencer™ Select |
Product Code | Brand | Purity | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Purity | Quantity | Price | Quantity & Availability | ||||
10714037
|
Ambion™
4427037 |
Standard | 1 nmol |
149.00 €
1nmol |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10782097
|
Ambion™
4392420 |
Standard | 5 nmol |
316.00 €
5nmol |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10584615
|
Ambion™
4392421 |
Standard | 20 nmol |
484.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10554045
|
Ambion™
4392422 |
Standard | 40 nmol |
564.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15551084
|
Ambion™
4444198 |
HPLC | 10 μmol |
14520.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15551064
|
Ambion™
4404012 |
In Vivo | 10 μmol |
15330.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry
- Silencer™ Select Pre-Designed siRNAs are guaranteed-to-silence based on their proven design
- Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening
- The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment.
The benefits of using Silencer™ Select siRNAs include:
- Potent – up to 100-fold more potent than currently available siRNAs and fewer off-target effects
- Most Specific – LNA™ chemical modifications reduce off-target effects by up to 90%
- Reliable – demonstrated improvements in consistency and reliability of phenotypic results
- Guaranteed – 100% guaranteed to silence, the best guarantee in the industry
Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.
Approximate Turnaround Times*
- Standard purification: 4 business days
- HPLC purification: 6 business days (20 and 40 nmol); 15 business days (250 nmol)
- In Vivo Ready: 15 business days (250 nmol)
Specifications
Silencer™ Select Pre-Designed siRNA | |
siRNA | |
Silencer™ Select |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.