man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Chromatography Columns and Cartridges
Column Packing
Macherey-Nagel™ Silica 60
Print
Macherey-Nagel™ Silica 60
77.25 € - 297.00 €
See promotional offers below
Specifications
Pore Size
60 Å
Type
Silica 60
Material
Silica
View More Specs
Products
2
Description
Specifications
Videos
SDS
Documents
Product Certifications
Products
2
Product Code
Brand
Quantity
Price
Quantity & Availability
Product Code
Brand
Quantity
Price
Quantity & Availability
10286281
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Macherey-Nagel™
815650.1
1 kg
77.25 €
1kg
In Stock
Log in
to see stock.
Add to basket
12730453
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Macherey-Nagel™
815650.5
5 kg
297.00 €
5kg
In Stock
Log in
to see stock.
Add to basket
Description
Description
Specifications
Specifications
Pore Size
60 Å
Type
Silica 60
Material
Silica
Videos
SDS
Documents
Documents
Product Certifications
Back to Top