missing translation for 'onlineSavingsMsg'
Learn More
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™ Product Code.: 10611573

Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™

Product Code. 10611573
500 g, Plastic Powder Jar
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Keg
Plastic Powder Jar
This item is not returnable. View return policy

Product Code. 10611573

Brand: Fisher Chemical S/0761/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Color Brown-Orange
Physical Form Solid
pH 1.5 to 5
Quantity 500 g
Packaging Plastic Powder Jar
IUPAC Name dioxosilane
Formula Weight 60.08g/mol
  • Fisher Chemical Silica gel, self indicating, 4 to 7 mesh (2.5mm to 6.0mm)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.