missing translation for 'onlineSavingsMsg'
Learn More

Silica gel 60, 0.019-0.037mm (400-600 mesh), Thermo Scientific Chemicals

Product Code. 11305517
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

AA4272918

Column chromatography with elevated pressureSilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Physical Form Powder
Color White
Quantity 50 g
Odor Odorless
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Chemical Name or Material Silica gel 60

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'