missing translation for 'onlineSavingsMsg'
Learn More

Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11309178
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg

Column chromatographySilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Physical Form Powder, -230+400 Mesh,S.A. 500-600m
Mesh Size 230-400 Mesh
Odor Odorless
Assay Percent Range 99.5% (metals basis)
Color White
Quantity 1 kg
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
IUPAC Name silanedione
Chemical Name or Material Silica gel 60

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'