missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, 60 to 200 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15568124
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    Suitable For Column Chromatography

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Material Silica Gel
Quantity 500 g
Mesh Size 60 to 200 Mesh
Packaging Wide Mouth Amber Glass Bottle
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Silica Gel
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'