Learn More
Silica Gel 60 Angstrom Pore Size, 60-200 Micron, for Column Chromatography, Fisher Chemical™
113.00 € - 138.00 €
Specifications
Packaging Plastic Powder Jar
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10254202
View Documents
Fisher Chemical
S/0694/53
500 g
113.00 €
500g
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
10346182
View Documents
Fisher Chemical
S/0694/60
1 kg
138.00 €
1kg
In Stock
to see stock.
Add to basket
Specifications
Plastic Powder Jar