missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica gel, for column chrom., ultrapure, 60-200μ, 60A, Thermo Scientific Chemicals
40.20 € - 1730.61 €
Chemical Identifiers
CAS | 7631-86-9 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | Silicon dioxide |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10078971
|
Thermo Scientific Acros
360062500 |
250 g | Plastic Bottle |
40.20 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10732643
|
Thermo Scientific Acros
360060010 |
1 kg | Plastic Bottle |
98.59 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914121
|
Thermo Scientific Acros
360060050 |
5 kg | Plastic Drum |
342.71 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16348237
|
Thermo Scientific Acros
360060300 |
30 kg | Plastic Bottle |
1730.61 €
30kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatographyChemical Identifiers
7631-86-9 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
O=[Si]=O |
O2Si | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
Silicon dioxide | |
CHEBI:30563 |
Specifications
White to Brown | |
6.5 to 7.5 (10% w/w suspension) | |
Silica Gel | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
Silicon dioxide | |
Solubility in water: insoluble. | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Silica gel, For column chromatography |
Powder and/or Chunks | |
7631-86-9 | |
O2Si | |
5.0% Max. (under size) 5.0% Max. (over size) | |
0.70 to 0.85mL/g (specific) | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08 |
Safety and Handling
Product Identifier
- Silica gel
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title