missing translation for 'onlineSavingsMsg'
Learn More
Silica gel desiccant, indicating, ACS, Thermo Scientific Chemicals Product Code.: 11343958

Silica gel desiccant, indicating, ACS, Thermo Scientific Chemicals

Product Code. 11343958
100 g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 11343958

Brand: Thermo Scientific Alfa Aesar 040381.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits.

Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Color Blue
Physical Form Granular
Density 2.17 to 2.66g/cm3
Material Silica Gel
Quantity 100 g
Odor Odorless
Solubility Information Soluble in alkalies. Insoluble in water.
Sensitivity Hygroscopic
Grade ACS
Chemical Name or Material Silica gel desiccant, For analysis ACS

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU07K024 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number040381.36,040381.A4,040381.22
Lot NumberQ18K031 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number040381.36,040381.A4,040381.22
Lot NumberT24J073 Certificate TypeCertificate of Analysis Date09/08/2023 Catalog Number040381.36,040381.A4,040381.22
Lot NumberX31I009 Certificate TypeCertificate of Analysis Date08/11/2022 Catalog Number040381.36,040381.A4,040381.22
Lot NumberN14I045 Certificate TypeCertificate of Analysis Date12/08/2022 Catalog Number040381.36,040381.A4,040381.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silica gel desiccant, indicating, ACS, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.