Learn More
Silica Gel Grade 923 Davisil™, for Column Chromatography, Fisher Chemical™
105.00 € - 445.00 €
Specifications
Packaging Plastic Powder Jar
pH 5.5 to 8
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10326232
View Documents
Fisher Chemical
S/0697/48
100 g
105.00 €
100g
10162682
View Documents
Fisher Chemical
S/0697/50
250 g
226.00 €
250g
10530423
View Documents
Fisher Chemical
S/0697/53
500 g
445.00 €
500g
Specifications
Plastic Powder Jar
5.5 to 8
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Silica gel
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Silica Gel Grade 923 Davisil™, for Column Chromatography, Fisher Chemical™