missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Gel, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
500g
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Material | Silica Gel |
Quantity | 2.5 kg |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Silica Gel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Silica Gel, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction