missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Gel PS Drying Agent, Honeywell Fluka™
With indicator
72.45 € - 336.51 €
Chemical Identifiers
CAS | 112926-00-8 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.083 |
MDL Number | MFCD00011232 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15627080
|
Honeywell-Fluka
78307-250G |
250 g | Poly Bottle | ||||||
15617080
|
Honeywell-Fluka
78307-1KG |
1 kg | Poly Bottle |
336.51 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
112926-00-8 | |
60.083 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
CHEBI:30563 | |
O=[Si]=O |
O2Si | |
MFCD00011232 | |
24261 | |
dioxosilane |
Specifications
112926-00-8 | |
O2Si | |
NONH for all modes of transport | |
O=[Si]=O | |
60.083 | |
CHEBI:30563 | |
Silica gel PS drying agent |
Silica Gel | |
MFCD00011232 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08g/mol |