missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel Rubin Drying Bags, Honeywell Fluka™

With indicator function, 1/3 TE, bag with ∼10 g, crystalline

53.95 € - 245.83 €

Specifications

Chemical Name or Material Silica gel Rubin drying bags
CAS 112926-00-8
Material Silica Gel
UN Number NONH for all modes of transport
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15667160
View Documents
Honeywell-Fluka
80148-250G
250 g Metal Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15696910
View Documents
Honeywell-Fluka
72811-250G
250 g Paper Poly Bag This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15657160
View Documents
Honeywell-Fluka
80148-1KG
1 kg Metal Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15686910
View Documents
Honeywell-Fluka
72811-1KG
1 kg Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Silica gel Rubin drying bags
112926-00-8
Silica Gel
NONH for all modes of transport
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : 231-545-4

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.