missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Standard SIL G Silica Layers on Glass Plates

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on glass plates come in a variety of plate sizes and layer thicknesses. Select a fluorescent indicator and choose from a variety of pack sizes.

Brand:  Macherey-Nagel™ 809121

 View more versions of this product

Product Code. 12770154

  • 230.00 € / Pack of 100
Estimated Shipment: 15-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on glass plates come in a variety of plate sizes and layer thicknesses. Select a fluorescent indicator and choose from a variety of pack sizes.

Glass TLC plates, SIL G/UV 254.
  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
Absorbent
  • Silica 60
  • specific surface (BET) ∼ 500 m2/g
  • Mean pore size 60 Å
  • Specific pore volume 0.75 mL/g,
  • Particle size 5–17 μm
  • Standard grade
Thickness
  • For analytical plates 0.25 mm
  • For preparative plates 0.5 and 1 mm
  • For 2 mm preparative layers a slightly coarser material is used.
Inicators
  • Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
  • Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
Binders
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications

Specifications

Silica 60
60 Å
Glass Plate
0.75 mL/g
0.25 mm
5 to 17 μm
100 Pk.
UV254 + UV366
∼500 m2/g
50 x 200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers