Learn More
Macherey-Nagel™ Silica Glass Plates
Brand: Macherey-Nagel™ 811054
This item is not returnable.
View return policy
Specifications
Silica 60 | |
60 Å | |
Glass Plate | |
∼500 m2/g | |
10 x 10 cm |
2 to 10 μm | |
25 Pk. | |
0.75 mL/g | |
0.20 mm |