missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicic Acid, Honeywell™ Fluka™
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Specifications
Specifications
Packaging | Poly Bottle |
Quantity | 1 kg |
Physical Form | Powder |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Silicic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction