missing translation for 'onlineSavingsMsg'
Learn More
Silicic acid hydrate, Thermo Scientific Chemicals Product Code.: 11387578

Silicic acid hydrate, Thermo Scientific Chemicals

Product Code. 11387578
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11387578

Brand: Thermo Scientific Alfa Aesar 033270.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silicic acid hydrate is used for studying neurodegenerative diseases due to its ability to inhibit aluminum uptake. It is utilized for the manufacture catalyst and catalyst carrier. It is involved in tungsten filament production as chemical reagent and flux. Further, it is used for oil and wax decoloring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1343-98-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym orthosilicic acid, monosilicic acid, tetrahydroxysilane, h4sio4, silicon tetrahydroxide, silicasol, kieselsaeure, si oh 4, unii-623b93yabh, orthosilicicacid
PubChem CID 14942
ChEBI CHEBI:26675
IUPAC Name silicic acid
SMILES O=[Si]=O

Specifications

Appearance White powder
Color White
Quantity 100 g
Physical Form -100 Mesh Powder
Formula Weight 60.09 (Anhydrous)
Chemical Name or Material Silicic acid hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN12L016 Certificate TypeCertificate of Analysis Date14/02/2025 Catalog Number033270,033270.36,033270.22
Lot NumberP15K012 Certificate TypeCertificate of Analysis Date21/03/2024 Catalog Number033270.36,033270.22
Lot NumberR08J033 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number033270.36,033270.22
Lot NumberN08J011 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number033270.36,033270.22
Lot NumberS30J048 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog Number033270,033270.36,033270.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicic acid hydrate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.