missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide, alpha-phase, 99.8% (metals basis), Thermo Scientific Chemicals

Surface Area 9-11 m2/g

74.67 € - 641.19 €

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
SMILES [C-]#[Si+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11379297
View Documents
Thermo Scientific Alfa Aesar
040155.22
100 g
74.67 €
100g
Please to purchase this item. Need a web account? Register with us today!
11399297
View Documents
Thermo Scientific Alfa Aesar
040155.36
500 g
205.02 €
500g
Please to purchase this item. Need a web account? Register with us today!
11389297
View Documents
Thermo Scientific Alfa Aesar
040155.A3
2 kg
641.19 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

409-21-2
40.10
HBMJWWWQQXIZIP-UHFFFAOYSA-N
9863
[C-]#[Si+]
CSi
MFCD00049531
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
CHEBI:29390

Specifications

2700°C
Black to Gray
99.8% (metals basis)
MFCD00049531
Soluble in molten sodium hydroxide,potassium hydroxide and in molten iron.
[C-]#[Si+]
40.10
CHEBI:29390
Odorless
409-21-2
Powder
CSi
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
HBMJWWWQQXIZIP-UHFFFAOYSA-N
methanidylidynesilylium
9863
40.1
Silicon carbide, alpha-phase
Videos
Safety and Handling

Safety and Handling

H351-H315-H319-H335: Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

missing translation for 'einecsNumber' : 206-991-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only