missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide, beta-phase, nanopowder, Thermo Scientific Chemicals

Product Code. 11435480
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

S.A. 70 to 90 m2/g

Silicon carbide can be used form very hard ceramics that are widely used in applications requiring high endurance, such as car brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide as light-emitting diodes (LEDs) and detectors in early radios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
SMILES [C-]#[Si+]

Specifications

Melting Point 2700°C
Color Black to Gray
Physical Form Powder
Quantity 5 g
Solubility Information Insoluble in water
IUPAC Name methanidylidynesilylium
Formula Weight 40.1
Odor Odorless
Chemical Name or Material Silicon carbide, beta-phase

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'