missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon(II) oxide, Optical Grade, 99.8% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 10097-28-6 |
---|---|
Molecular Formula | OSi |
Molecular Weight (g/mol) | 44.084 |
MDL Number | MFCD00151536 |
InChI Key | LIVNPJMFVYWSIS-UHFFFAOYSA-N |
Synonym | silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene |
PubChem CID | 66241 |
ChEBI | CHEBI:30588 |
IUPAC Name | oxoniumylidynesilanide |
SMILES | [O+]#[Si-] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10097-28-6 | |
44.084 | |
LIVNPJMFVYWSIS-UHFFFAOYSA-N | |
66241 | |
oxoniumylidynesilanide |
OSi | |
MFCD00151536 | |
silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene | |
CHEBI:30588 | |
[O+]#[Si-] |
Specifications
1880°C | |
10097-28-6 | |
Powder | |
OSi | |
silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene | |
[O+]#[Si-] | |
44.084 | |
CHEBI:30588 | |
Optical | |
2.13 |
>1702°C | |
Black | |
99.8% (metals basis) | |
MFCD00151536 | |
LIVNPJMFVYWSIS-UHFFFAOYSA-N | |
oxoniumylidynesilanide | |
66241 | |
44.09 | |
Odorless | |
Silicon(II) oxide |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 233-232-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only