missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) iodide, 99% (metals basis), Thermo Scientific Chemicals

Light, Air and Moisture Sensitive

43.00 € - 430.14 €

Chemical Identifiers

CAS 13465-84-4
Molecular Formula I4Si
Molecular Weight (g/mol) 535.70
MDL Number MFCD00054041
InChI Key CFTHARXEQHJSEH-UHFFFAOYSA-N
Synonym silicon tetraiodide, silane, tetraiodo, silicon iv iodide, periodosilane, tetraiodosilicon, acmc-1bu37, silicon tetraiodide trace metals basis, silicon iv iodide trace metals basis 1g
PubChem CID 83498
IUPAC Name tetraiodosilane
SMILES I[Si](I)(I)I
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364969
View Documents
Thermo Scientific Alfa Aesar
087303.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11354969
View Documents
Thermo Scientific Alfa Aesar
087303.06
5 g
145.73 €
5g
Please to purchase this item. Need a web account? Register with us today!
11344969
View Documents
Thermo Scientific Alfa Aesar
087303.14
25 g
430.14 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Silicon(IV) iodide is used as a precursor to prepare silicon amides. It finds application in the manufacture and etching of silicon in microelectronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13465-84-4
535.70
CFTHARXEQHJSEH-UHFFFAOYSA-N
83498
I[Si](I)(I)I
I4Si
MFCD00054041
silicon tetraiodide, silane, tetraiodo, silicon iv iodide, periodosilane, tetraiodosilicon, acmc-1bu37, silicon tetraiodide trace metals basis, silicon iv iodide trace metals basis 1g
tetraiodosilane

Specifications

13465-84-4
Crystals
I4Si
silicon tetraiodide, silane, tetraiodo, silicon iv iodide, periodosilane, tetraiodosilicon, acmc-1bu37, silicon tetraiodide trace metals basis, silicon iv iodide trace metals basis 1g
I[Si](I)(I)I
535.70
535.7
White
99% (metals basis)
MFCD00054041
CFTHARXEQHJSEH-UHFFFAOYSA-N
tetraiodosilane
83498
Silicon(IV) iodide
Videos
Safety and Handling

Safety and Handling

H300-H314-H318: Fatal if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage.

missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II

missing translation for 'einecsNumber' : 236-706-2

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.